Order Entry
Canada
Orders LinkContactUsLinkComponent
 

ScienceLine pH Combination Electrodes, SI Analytics

Electrode, H 61, pH glass H, 12 mm, 170 mm, Plug head, without cable, KCl 3 mol/L, Glass, +10...+100 °C, 0 - 14, Platinum
  97041-980
 :  285100207
ScienceLine pH Combination Electrodes, SI Analytics
  97041-980
 :  
ScienceLine pH Combination Electrodes, SI Analytics
ScienceLine pH Combination Electrodes, SI Analytics

Electrode, H 61, pH glass H, 12 mm, 170 mm, Plug head, without cable, KCl 3 mol/L, Glass, +10...+100 °C, 0 - 14, Platinum

 
 

pH combination electrodes are available with plug head and no cable or BNC plug and fixed cable of one meter length.

 : Connection cables for electrodes with plug heads are sold separately, contact your Avantor representative for more information.

 
  • Ø:
    12 mm
  • Temperature range:
    +10...+100 °C
  • Type:
    H 61
  • Description:
    Electrode
  • Length:
    170 mm
  • Material:
    pH glass H
  • pH range:
    0 - 14
  • Body material:
    Glass
  • Junction:
    Platinum
  • Connector:
    Plug head, without cable
  • Electrolyte:
    KCl 3 mol/L
  • Cat. No.:
    97041-980
  • Shape:
    Sphere, except type L 8280 (Dome)
  • Reference system:
    Silamid®, except type N 6480 Eis (Ag/AgCl-reference system)