Orders LinkContactUsLinkComponent
Pegboard Mounting Assemblies, Finished Back, Interdyne
Pegboard mounting assembly, Stainless steel 304, 30×30"
Catalog # 97034-234
Supplier:   Interdyne
 :  FB-3030
Pegboard Mounting Assemblies, Finished Back, Interdyne
Catalog # 97034-234
Supplier:  Interdyne
Pegboard Mounting Assemblies, Finished Back, Interdyne
Pegboard mounting assembly, Stainless steel 304, 30×30"
Price
$681.07
Log in for availability
Each
Product Details & Documents
The assembly is made from 20 gauge stainless steel with ³/₄" turned edge.
Specifications
  • Description:
    Pegboard mounting assembly
  • Material:
    Stainless steel 304
  • Ext. dimensions:
    30×30"