Pegboard Mounting Assemblies, Finished Back, Interdyne
Pegboard mounting assembly, Stainless steel 304, 24×30"
Catalog # 97034-232
Supplier: Interdyne
: FB-2430
Jump to:
Product Details & Documents
The assembly is made from 20 gauge stainless steel with ³/₄" turned edge.
Specifications
- Description:Pegboard mounting assembly
- Material:Stainless steel 304
- Ext. dimensions:24×30"