Order Entry
Canada
Orders LinkContactUsLinkComponent
Cabinet Accessories, Justrite®

Cabinet Accessories, Justrite®

SpillSlope™ Steel Shelf for Safety Cabinets, 20 gal. Wall Mount cabinets, 39⅜", 8⅝", 44", 6 lbs.
Catalog # 97001-316
Supplier:   Justrite
Supplier #:  29938
Cabinet Accessories, Justrite®
Catalog # 97001-316
Supplier:  Justrite
Cabinet Accessories, Justrite®
Cabinet Accessories, Justrite®

SpillSlope™ Steel Shelf for Safety Cabinets, 20 gal. Wall Mount cabinets, 39⅜", 8⅝", 44", 6 lbs.

Price
$152.75
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents

Cabinet accessories offer an opportunity to expand storage and add new convenience to existing cabinets, or customize new cabinets with additional shelves to meet specific workflow needs.

  • Expand storage
  • Add new convenience
  • Customize new cabinets

Polyethylene trays offer additional protection and have a slightly raised curved edge to contain spills

SpillSlope™ galvanized steel shelves hold up to 159 kg (350 lbs.) and adjust on 5.7 cm (21/4") increments for versatile storage.

They may also be used in the bottom sill area to catch incidental spills and leaks.

Documents

Specifications
  • Depth:
    8⅝"
  • Width:
    39⅜"
  • Description:
    SpillSlope™ Steel Shelf for Safety Cabinets
  • Height:
    44"
  • Shipping weight:
    6 lbs.
  • For use with:
    20 gal. Wall Mount cabinets
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.