Order Entry
Canada
Orders LinkContactUsLinkComponent
Scissors, Strong, Excelta Corp®

Scissors, Strong, Excelta Corp®

Scissors, kevlar, 3 star quality, 5.5" (137.5 mm), Stainless steel AISI 440, 44 mm (1¾"), Straight
Catalog # 94024-214
Supplier:   Excelta
Supplier #:  299P
Scissors, Strong, Excelta Corp®
Catalog # 94024-214
Supplier:  Excelta
Scissors, Strong, Excelta Corp®
Scissors, Strong, Excelta Corp®

Scissors, kevlar, 3 star quality, 5.5" (137.5 mm), Stainless steel AISI 440, 44 mm (1¾"), Straight

Price
$83.75
Log in for availability
Each
Product Details & Documents

Heavy duty applications require large strong scissors, and these fit that bill.

  • Designed for heavy duty applications
  • Serrated outside edges

These heavy duty general purpose scissors have serrated outside blade edges. They are set up for heavy duty cutting.

Specifications
  • Description:
    Scissors, kevlar, 3 star quality
  • Length:
    5.5" (137.5 mm)
  • Material:
    Stainless steel AISI 440
  • Curvature:
    Straight
  • Blade length:
    44 mm (1¾")