VWR® Flat Mops
Product 68400-408 is similar to 94023-760 but manufacturer and specifications may vary.
Product Details & Documents
These brick-style, flat sponge mop heads are excellent for applying disinfectant in pharmaceutical production or aseptic processing applications.
- Autoclavable
- Easily cleans large surfaces
- Heavy-duty construction offers additional resistance to textured floors and aggressive cleaning methods
Fully padded mop heads can be laundered, autoclaved, and reused several times.
Ideal for cleaning large surface areas as well as reaching shelving, doorjambs, and corners.
Mop heads feature heavy-duty construction with PolyTex™ textured polyester material, PolyTex™ material combined with MegaTex™ textured non-woven polyamide fabric strip, or PolyTex™ material combined with microfiber fabric strip. PolyTex™/MegaTex™ mop heads offer additional resistance to textured floors and more aggressive action for spills and heavy soils. PolyTex™/microfiber mop heads allow faster pick up of moisture in standard clean/disinfect applications such as hospitals and animal labs.
Ordering information: All components must be purchased separately. An electropolished stainless steel quick-disconnect adapter is required to attach mop head to an electropolished stainless steel extension handle. For adapters and handles, contact your Avantor representative.
Documents
Certificates
Search by Lot NumberVWR Cleaning Essentials for Controlled and Critical Environments
Product Literature
Accessories
Product Information
- Catalog #MaterialLengthAvailabilityPrice
- Stainless steel quick disconnect extendable handleStainless steel quick disconnect extendable handle68400-164Stainless steel 304, electropolishedExtendable from 96.5 - 182.9 cm (38 - 72")Log in for availabilityEach - $464.98