Orders LinkContactUsLinkComponent
VWR® Flat Mops
VWR® Flat Mops
PolyTex™/MegaTex™ mop heads, 36 cm (14")
Catalog # 94023-754
VWR® Flat Mops
Catalog # 94023-754
Supplier:  
VWR® Flat Mops
VWR® Flat Mops
PolyTex™/MegaTex™ mop heads, 36 cm (14")
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
SLIMLINE MOP ADAPTER 14IN Catalog #11310-598
Product 11310-598 is similar to 94023-754 but manufacturer and specifications may vary.
Product Details & Documents
These brick-style, flat sponge mop heads are excellent for applying disinfectant in pharmaceutical production or aseptic processing applications.
  • Autoclavable
  • Easily cleans large surfaces
  • Heavy-duty construction offers additional resistance to textured floors and aggressive cleaning methods
Fully padded mop heads can be laundered, autoclaved, and reused several times.
Ideal for cleaning large surface areas as well as reaching shelving, doorjambs, and corners.
Mop heads feature heavy-duty construction with PolyTex™ textured polyester material, PolyTex™ material combined with MegaTex™ textured non-woven polyamide fabric strip, or PolyTex™ material combined with microfiber fabric strip. PolyTex™/MegaTex™ mop heads offer additional resistance to textured floors and more aggressive action for spills and heavy soils. PolyTex™/microfiber mop heads allow faster pick up of moisture in standard clean/disinfect applications such as hospitals and animal labs.
Ordering information: All components must be purchased separately. An electropolished stainless steel quick-disconnect adapter is required to attach mop head to an electropolished stainless steel extension handle. For adapters and handles, contact your Avantor representative.
Specifications
  • Description:
    PolyTex™/MegaTex™ mop heads
  • Size:
    36 cm (14")
Accessories
Product Information
  • Catalog #
    Material
    Length
    Availability
    Price
  • Stainless steel quick disconnect extendable handle
    Stainless steel quick disconnect extendable handle
    68400-164
    Stainless steel 304, electropolished
    Extendable from 96.5 - 182.9 cm (38 - 72")
    Log in for availability
    Each - $464.98