Order Entry
Canada
Orders LinkContactUsLinkComponent
NMC Valve Wheel Lockouts

NMC Valve Wheel Lockouts

Fits 2.54 cm (1") To 6.35 cm (2.5") Dia.
  94022-904
Supplier #:  VL02R
NMC Valve Wheel Lockouts
  94022-904
NMC Valve Wheel Lockouts
NMC Valve Wheel Lockouts

Fits 2.54 cm (1") To 6.35 cm (2.5") Dia.

 
$98.92
   
Each
Minimum order quantity is 1
 

Lockouts consist of two flattened half moons that completely cover the valve wheels from 1" to 14" in diameter

Made from strong Polypro plastic, fully dielectric. Highly resistant to cracking and abrasion. Resistant to extreme temperature changes 50⁰F

 
  • Description:
    Fits 2.54 cm (1") To 6.35 cm (2.5") Dia.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.