Order Entry
Canada
Orders LinkContactUsLinkComponent
 

MasterLock® Lockout Padlocks, NMC

1.91 cm (3/4"), White
Catalog # 94017-588
 :  MPSW
MasterLock® Lockout Padlocks, NMC
Catalog # 94017-588
MasterLock® Lockout Padlocks, NMC
MasterLock® Lockout Padlocks, NMC

1.91 cm (3/4"), White

Price
88,93 $
Log in for availability
Each
 
Product Details & Documents

Over 10,000 unique key codes, 4 pin tumbler cylinders

Heavy duty laminated steel case constructed with hardened shackles. Full keying capability for most commercial applications. Locks are keyed differently. Two keys included.

Specifications
  • Color:
    White
  • Shackle size:
    1.91 cm (3/4")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.