MasterLock® Lockout Padlocks, NMC
3.81 cm (11/2"), Red
Catalog # 94017-574
Supplier: ACCUFORM MANUFACTURING, INC
: MPLR
Jump to:
Product Details & Documents
Over 10,000 unique key codes, 4 pin tumbler cylinders
Heavy duty laminated steel case constructed with hardened shackles. Full keying capability for most commercial applications. Locks are keyed differently. Two keys included.
Specifications
- Color:Red
- Shackle size:3.81 cm (11/2")



