Orders LinkContactUsLinkComponent
NMC Lockout Stations
NMC Lockout Stations
Lockout Center Equipped
Catalog # 94016-426
NMC Lockout Stations
Catalog # 94016-426
Supplier:  
NMC Lockout Stations
NMC Lockout Stations
Lockout Center Equipped
Discontinued
Product Details & Documents
High visibility stations facilitate proper and convenient lockouts
Heavy duty plexiglass board features replaceable hooks for locks and clear pockets for tags. Features lightweight Xenoy® lock bodies. Available in 48.26 x 60.96 cm (19 x 24") and 20.32 x 30.48 cm (8 x 12").
Specifications
  • Description:
    Lockout Center Equipped