NMC Group Lock Box
Group Lockbox, 15.24 x 26.04 x 11.43 cm (6 x 101/4 x 41/4")
Catalog # 94014-974
Supplier: ACCUFORM MANUFACTURING, INC
Supplier #: GLB02
Jump to:
Product Details & Documents
Use for work team situations where there are multiple workers and energy sources all keys are stored in box
Group leader locks box until job is completed and members are cleared. Box is made from a heavy steal construction.
Specifications
- Description:Group Lockbox
- Dimensions:15.24 x 26.04 x 11.43 cm (6 x 101/4 x 41/4")