Polypropylene Buckets, Contec®
PP bucket, Grey, 24.6 L (6.5 gal.)
: UBKT0003
Contec® bucket systems are available in a variety of size options for small and large critical environments. Replacement buckets are available for the polypropylene and stainless systems.
- Replacement buckets for polypropylene and stainless bucket systems
- For use with 3.9, 5, 6.5, and 7 gal. bucket systems
- Autoclavable
- Available in a variety of colors
These buckets are autoclavable and work with a variety of disinfectants and solvents. Wringer options are available based on bucket size.
- Capacity:24.6 L (6.5 gal.)
- Color:Grey
- Description:PP bucket
Accessories
- Description
- DescriptionColorCapacity
- DescriptionMaterialFor Use With
- 89198-854Flat Mop WringerPPVertiKlean® MAX™ and QuickTask™ Mops (25 l (6.5 gal.) Buckets)Case of 1 - $130.72



