Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Axygen® Gel Loading Tips, Corning

Round, O.D. 0.057 cm (0.022"), 200 µl, Clear, 200 Tips/Rack
  89179-806
 :   Corning
 :  TGL-200RD-57-R
Axygen® Gel Loading Tips, Corning
  89179-806
 :  Corning
Axygen® Gel Loading Tips, Corning
Axygen® Gel Loading Tips, Corning

Round, O.D. 0.057 cm (0.022"), 200 µl, Clear, 200 Tips/Rack

 
$140.25
 
   
Case of 400
 

Axygen brand gel loading tips have a universal fit for reseach grade pipettors.

They have a convenient graduation mark at 5 µl for quick volume check. The 0.17 mm tips are for loading 0.2 mm thick gels.

 
  • Volume:
    200 µl
  • Tip color:
    Clear
  • Packed:
    200 Tips/Rack
  • Tip style:
    Round, O.D. 0.057 cm (0.022")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.