Order Entry
Canada
Orders LinkContactUsLinkComponent
Sample Vials, Clear Borosilicate Glass, Screw-Thread

Sample Vials, Clear Borosilicate Glass, Screw-Thread

Vials with treatment, cleaned and certified for volatiles, 20.00 ml, 27.25×57.5 mm, Clear, Yes, 24-400
Catalog # 89176-854
Supplier:   Qorpak
Supplier #:  GLC-01015
Sample Vials, Clear Borosilicate Glass, Screw-Thread
Catalog # 89176-854
Supplier:  Qorpak
Sample Vials, Clear Borosilicate Glass, Screw-Thread
Sample Vials, Clear Borosilicate Glass, Screw-Thread

Vials with treatment, cleaned and certified for volatiles, 20.00 ml, 27.25×57.5 mm, Clear, Yes, 24-400

Price
$482.19
New Customer Promotional Price
Log in for availability
Case of 72
Product Details & Documents

Clear, flint screw-thread sample vials offer maximum visibility.

  • Available with a variety of caps, or as vials only, to meet specific requirements
  • Pre-cleaned options save time and money

These general-purpose glass vials are made of Type I Borosilicate glass, providing exceptional resistance to heat shock and chemical leaching.

Specifications
  • Cap included:
    Yes
  • Cap Size:
    24-400
  • Capacity:
    20.00 ml
  • Color:
    Clear
  • Description:
    Vials with treatment, cleaned and certified for volatiles
  • Diameter ×H:
    27.25×57.5 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.