Order Entry
Canada
Orders LinkContactUsLinkComponent
Protector® Combination Glove Box, Labconco®

Protector® Combination Glove Box, Labconco®

ULPA Filtered, Double Width, 208/230 V, 60 Hz, (US), Stainless steel
Catalog # 89175-400
Supplier:   Labconco
Supplier #:  5080182
Protector® Combination Glove Box, Labconco®
Catalog # 89175-400
Supplier:  Labconco
Protector® Combination Glove Box, Labconco®
Protector® Combination Glove Box, Labconco®

ULPA Filtered, Double Width, 208/230 V, 60 Hz, (US), Stainless steel

Price
$100,230.00
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Protector® Combination Glove Boxes provide inlet and outlet HEPA or ULPA filtration and a leak-tight physical barrier to protect the operator from exposure to potentially hazardous materials and perform work with contamination-sensitive materials under vacuum or in an inert environment. Protector Combination Glove Boxes feature two internal valves to convert the glove box from a flow-through filtered box to an inert atmosphere glove box

All Protector Combination Glove Boxes are compatible with the new AtmosPure Re-Gen Gas Purifier accessory, which has the capacity to remove up to five liters of oxygen (13,500 ppm) and 660 grams of moisture (18 million ppm) at standard temperature and pressure (STP).

Accessories information: Glove box interior shelves kit provides shelf space across the back wall. Three stainless steel storage shelves are attached to epoxy-coated steel upright supports. Shelves have adjustable height. Installation hardware and instructions are included.

Specifications
  • Description:
    ULPA Filtered, Double Width
  • Material:
    Stainless steel
  • Electrical:
    208/230 V, 60 Hz, (US)
  • Certification:
    UL, ETL, CAN/CSA
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Glove Box Interior Shelves Kit, 76.2W x 15.2D cm (30 x 6")
    Glove Box Interior Shelves Kit, 76.2W x 15.2D cm (30 x 6")
    32940-080
    Log in for availability
    Each - $1,120.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.