Order Entry
Canada
Orders LinkContactUsLinkComponent
General Printer Labels, Brady

General Printer Labels, Brady

Printer labels, B-569, Polyester, Yellow, BBP®31, BBP®33, 4", 30.48 m (100 ft.)
Catalog # 89174-934
Supplier:   Brady Worldwide
Supplier #:  117990
General Printer Labels, Brady
Catalog # 89174-934
Supplier:  Brady Worldwide
General Printer Labels, Brady
General Printer Labels, Brady

Printer labels, B-569, Polyester, Yellow, BBP®31, BBP®33, 4", 30.48 m (100 ft.)

Price
$952.99
New Customer Promotional Price
Log in for availability
Roll of 1
Product Details & Documents

General labels for straws, petri dishes, bottles, flasks, microplates, bags, and more.

Tamper evident labels and tapes: B-438 metalized polyester, permanent adhesive designed to leave checkboard footprint if removed.

Specifications
  • For Use With:
    BBP®31, BBP®33
  • Color:
    Yellow
  • Description:
    Printer labels, B-569
  • Material:
    Polyester
  • Roll length:
    30.48 m (100 ft.)
  • Width:
    4"
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.