Order Entry
Canada
Orders LinkContactUsLinkComponent
 

General Printer Labels, Brady

Printer labels, B-569, Polyester, Blue, BBP®31, BBP®33, 4", 30.48 m (100 ft.)
Catalog # 89174-884
Supplier:   Brady Worldwide
 :  117933
General Printer Labels, Brady
Catalog # 89174-884
Supplier:  Brady Worldwide
General Printer Labels, Brady
General Printer Labels, Brady

Printer labels, B-569, Polyester, Blue, BBP®31, BBP®33, 4", 30.48 m (100 ft.)

Price
$952.99
New Customer Promotional Price
Log in for availability
Roll of 1
Product Details & Documents

General labels for straws, petri dishes, bottles, flasks, microplates, bags, and more.

Tamper evident labels and tapes: B-438 metalized polyester, permanent adhesive designed to leave checkboard footprint if removed.

Specifications
  • For Use With:
    BBP®31, BBP®33
  • Color:
    Blue
  • Description:
    Printer labels, B-569
  • Material:
    Polyester
  • Roll length:
    30.48 m (100 ft.)
  • Width:
    4"