Order Entry
Canada
Orders LinkContactUsLinkComponent
BA 410E Compound Upright Microscope, Motic Instruments

BA 410E Compound Upright Microscope, Motic Instruments

CCIS EC-H PL 20X/ 0.40
Catalog # 89168-460
BA 410E Compound Upright Microscope, Motic Instruments
Catalog # 89168-460
Supplier:  
BA 410E Compound Upright Microscope, Motic Instruments
BA 410E Compound Upright Microscope, Motic Instruments

CCIS EC-H PL 20X/ 0.40

Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
CCIS EC-H PL 20X/0.40 OBJ Catalog #CA89168-460

Product CA89168-460 is a replacement for 89168-460 and has the same packaging and specifications, but may be a different quantity.

Product Details & Documents

The BA410 Elite is the flagship of Motic's biological microscope series, and is intended for a wide array of laboratory and research applications

  • Binocular head Siedentopf type 30° inclined, 360° rotating (binocular model)
  • Trinocular head Siedentopf type 30° inclined, 360° rotating [light split 20/80] (trinocular model)
  • Widefield high eyepoint eyepiece WFPL 10×/22 mm with diopter adjustment on both eyepieces, with rubber eyecup (paired)
  • Reversed sextuple nosepiece
  • CCIS Plan Achromat objectives EC-H PL4×, EC-H PL10×, EC-H PL40× S, EC-H PL100X S-oil
  • Coaxial coarse and fine focusing adjustment
  • Built in low position rackless stage with right hand control
  • Achromat swing-out condenser N.A. 0.90 with iris diaphragm
  • Koehler illumination 3W LED with external lamp house and intensity control
  • Universal power supply 100 - 240 V UL
  • Immersion oil (5 ml), power cord, allen hexagonal key, vinyl dust cover

Standard with the BA410 Elite series is Motic’s Enhanced Contrast (EC-H) plan achromatic lenses. These are easily upgraded to Motic’s plan fluar or plan apochromatic lenses for further enhanced image fidelity. A large rackless mechanical stage featuring smooth, ball-bearing drive motion, and 1mm graduated scale allows for precise, repeatable positioning of specimen slides. Robust design and exceptional upgradeability ensure that the BA410 Elite will be a work-horse for years to come.

Modular by design, the BA410 Elite is upgradeable with phase contrast, polarizing, epi-fluorescence, and discussion/teaching devices available to further increase the utility of this microscope. The professional focusing, swing-out Abbe condenser offers focused, consistent lighting across the entire magnification range providing exact, publication quality images when coupled with a Moticam camera.

Ordering information: Microscope is supplied with Micrometer reticles, phase contrast accessories, polarizing accessories, Darkfield accessories, and Epi-fluorescence attachments. Contact your Avantor representative to learn more about this and other Motic solutions. All Motic products come with a one-year electrical and five-year mechanical warranty.

Specifications
  • Description:
    CCIS EC-H PL 20X/ 0.40
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Left Armrest
    Left Armrest
    CA19000-554
    Log in for availability
    Each - 41,04 $

Product Information

  • Catalog #
    Availability
    Price
  • Right armrest
    Right armrest
    CA19000-556
    Log in for availability
    Each - 40,59 $

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.