Corning® Disposable Spinner Flasks, Corning
Corning® 500 mL Disposable Spinner Flask with 100 mm Top Cap and 2 Angled Sidearms with Vent Caps, 500 ml, 45 mm, 87.3×203.2 mm, Plain
Supplier: Corning
Supplier #: 3578
Jump to:
Product Details & Documents
The Corning® disposable spinner flask system comes ready-to-use with paddle and integrated magnet, eliminating the need for time-consuming assembly or cleaning and reassembly.
- Paddle size and height is optimised for each vessel size
- Integrated magnet provides smooth, even rotation at required speeds on slow-speed stirrers
- Heat build-up in the vessel is reduced by means of a specially designed flange that raised the vessel off the stir-plate surface for the 125 and 500 ml flasks only
Molded from virgin polystyrene and gamma-irradiated, each spinner flask system assures a clean and sterile unit that is nonpyrogenic. No more concerns with detergent residues or contamination.
Made of USP Class VI and ISO 10993-18 polystyrene, the vessel is comparable to conventional glass spinner flasks for growth of suspension cell lines and any attachment-dependent cultures using microcarrier beads.
Specifications
- Capacity:500 ml
- Description:Corning® 500 mL Disposable Spinner Flask with 100 mm Top Cap and 2 Angled Sidearms with Vent Caps
- Style:Plain
- Diameter ×H:87.3×203.2 mm
- Sidearm neck O.D.:45 mm
Accessories
Product Information
- Catalog #MaterialSterilityAvailabilityPrice
- Vent capVent cap89089-914High Density Polyethylene, orangeSterileLog in for availabilityCase of 4 - $68.85
Product Information
- Catalog #MaterialSterilityAvailabilityPrice
- Aseptic transfer cap
Product Information
- Catalog #Fits FlaskAvailabilityPrice
- Modified Impeller Assembly for Magnetically-Driven 36L Corning® Proculture® Spinner Flask, ReusableModified Impeller Assembly for Magnetically-Driven 36L Corning® Proculture® Spinner Flask, Reusable89089-95036L Corning® Proculture® spinner flask (22877-088, 22877-098, 22877-108, 22877-132, 22877-138, 22877-142)Log in for availabilityCase of 1 - $1,450.00
Product Information
- Catalog #MaterialSterilityAvailabilityPrice
- Aseptic transfer capAseptic transfer cap89089-912High Density Polyethylene, orangeSterileLog in for availabilityCase of 2 - $689.35
Product Information
- Catalog #MaterialSterilityAvailabilityPrice
- Aseptic transfer capAseptic transfer cap89089-908High Density Polyethylene, orangeSterileLog in for availabilityCase of 2 - $641.75



