Order Entry
Canada
Orders LinkContactUsLinkComponent
C75 ≥98%

C75 ≥98%

5 mg
Catalog # 89147-566
Supplier:   Enzo Life Sciences
Supplier #:  270-286-M005
C75 ≥98%
Catalog # 89147-566
Supplier:  Enzo Life Sciences
CAS Number:  191282-48-1
C75 ≥98%
C75 ≥98%

5 mg

CAS Number:  191282-48-1
Price
$624.10
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Fatty acid synthase inhibitor

Inhibitor of fatty acid synthase (FAS) reducing food intake and body weight in mice. Exhibits irreversible slow-binding biphasic inactivation of fatty acid synthase. Downregulates neuropeptide Y and Agouti-related protein expression. Has been proposed to activate CPT-1 activity in liver and adipose tissue, leading to increased fatty acid oxidation and energy production. Shows significant in vivo antitumor activity in human breast cancer cells. Suppresses DNA replication and induces apoptosis. FAS inhibition by C75 leads to a dramatic accumulation of the CDK inhibitor p27KIP1 from cytosol to cell nuclei.

Synthetic.

Synonyms:  trans-4-Methylene-2-octyl-5-oxotetrahydrofuran-3-carboxylic acid
  • CAS Number:191282-48-1
  • Formula:C₁₄H₂₂O₄
  • MDL Number:MFCD03265452
  • Molecular Weight:254.3221 g/mol
Specifications

Specifications

  • Size:
    5 mg

Specifications