Orders LinkContactUsLinkComponent
Reactive Dye Conjugates, Biotium
Reactive Dye Conjugates, Biotium
Aminooxy-5(6)-ROX, 568/595 nm, 5 mg
Catalog # 89139-708
Supplier:   Biotium
Supplier #:  90104
Reactive Dye Conjugates, Biotium
Catalog # 89139-708
Supplier:  Biotium
CAS Number:  
Reactive Dye Conjugates, Biotium
Aminooxy-5(6)-ROX, 568/595 nm, 5 mg
CAS Number:  
Price
$399.25
Log in for availability
Each (5mg)
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Reactive dyes containing an amine functional group.
  • Aminooxy conjugates
  • Cadaverine conjugates
  • Can be used to label abasic sites in damaged DNA
Our aminooxy reagents such as aminooxy-5(6)-FAM react with aldehydes or ketones to form a stable hydrazone compound under mild conditions
Dye conjugates to PEO3-amine have a long water-soluble PEG spacer that separates the fluorescent dye and the amine functional group. These dyes can be coupled to activated carboxy groups or sulfonyl chlorides.
Cadaverine conjugates can be coupled to activated carboxy groups or sulfonyl chlorides.
Similar to ARP, aminooxy-5(6)-FAM can be used to label abasic sites in damaged DNA.
Specifications
Specifications
  • Description:
    Aminooxy-5(6)-ROX
  • Size:
    5 mg
  • Ex/em:
    568/595 nm