Orders LinkContactUsLinkComponent
Colorimetric and Fluorogenic Enzyme Substrates, Biotium
Colorimetric and Fluorogenic Enzyme Substrates, Biotium
Chromogenic Phosphatase Substrate Sampler Kit, 1 kit
Catalog # 89138-240
Supplier:   Biotium
Supplier #:  10022
Colorimetric and Fluorogenic Enzyme Substrates, Biotium
Catalog # 89138-240
Supplier:  Biotium
CAS Number:  
Colorimetric and Fluorogenic Enzyme Substrates, Biotium
Chromogenic Phosphatase Substrate Sampler Kit, 1 kit
CAS Number:  
Price
$442.19
Log in for availability
Each
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Popular chromogenic and fluorogenic substrates for phosphatases, peroxidases, and other enzymes.
  • Wide selection of popular substrates for alkaline phosphatase, horseradish peroxidase, beta-galactosidase, and other common enzymes
  • Chromogenic substrates for absorbance-based detection or light microscopy
  • Fluorogenic substrates for highly sensitive fluorescence-based ELISA
  • Substrate sampler kits available
Biotium offers a number of popular substrates for alkaline phosphatase, beta-galactosidase, and horseradish peroxidase which are widely used for chromogenic or fluorogenic ELISA, immunohistochemistry, or blotting applications.
Beta-glucuronidase substrates are widely used to detect E. coli bacterial contamination in food and water, and also in reporter gene assays in plants.
X-beta-D-cellobioside is a substrate for beta-cellobiosidase.
ABTS is a registered trademark of Roche. Amplex Red is a registered trademark of Thermo Fisher Scientific.
Specifications
Specifications
  • Description:
    Chromogenic Phosphatase Substrate Sampler Kit
  • Size:
    1 kit
Specifications
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.