Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Canada
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
MOPS MICROPAD DRY 18IN CS6
Catalog # 89134-282
Supplier:
Vileda Professional - FHP
Expand image
MOPS MICROPAD DRY 18IN CS6
Catalog # 89134-282
Supplier:
Vileda Professional - FHP
Supplier Number:
131602
Frequently Bought Together
VWR® Disposable Serological Pipettes
Supplier: VWR International
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
VWR® Universal Pipette Tips
Supplier: VWR International