Order Entry
Canada
Orders LinkContactUsLinkComponent
 

HBIO BTXpress Solution, BTX

BTXpress solution kit, 40 in 4 mm gap cuvettes
Catalog # 89130-550
Supplier:   Harvard Apparatus
 :  45-0807
HBIO BTXpress Solution, BTX
Catalog # 89130-550
Supplier:  Harvard Apparatus
CAS Number:  
HBIO BTXpress Solution, BTX
HBIO BTXpress Solution, BTX

BTXpress solution kit, 40 in 4 mm gap cuvettes

CAS Number:  
Price
$1,665.47
Log in for availability
Each
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Single buffer solution developed to quickly and efficiently deliver genes into mammalian cells that were previously considered 'hard to transfect' by chemical and other non-viral methods.

  • High efficiency transfection of difficult cell lines
  • High cell viability
  • Low toxicity resulting in improved cell viability
  • Single buffer for all cell types
  • One buffer used in place of standard electroporation buffers for all mammalian cell types

BTXpress Electroporation Solution is the first electroporation reagent that meets all of your high performance transfection needs without sacrificing control over your experiment or your budget. It offers increased numbers of transfections per kit compared to other commercial alternatives, providing higher value to you.

As a universal solution, the BTXpress Electroporation Solution can be used in other electroporators, including the Amaxa™, to deliver similar results without the typical high cost associated with these buffer kits. The BTXpress Electroporation Solution is offered as a kit including the BTX Cuvettes Plus with transfer pipettes or as a buffer alone.

 : Trial Sample Kit includes 2 ml volume solution with five 2 mm gap cuvettes and five 4 mm gap cuvettes. The 2 ml volume is the solution only and it requires cuvettes. The The 5 ml volume is the solution only for up to 50 reactions. It requires twenty 4 mm gap cuvettes or fifty 2 mm gap cuvettes. The 10 ml volume is the solution only for up to 100 reactions. It requires forty 4 mm gap cuvettes or one hundred 2 mm gap cuvettes. The 20 Reaction Kit includes 5 ml volume solution and twenty 4 mm gap cuvettes. The 40 Reaction Kit includes 5 ml volume solution and forty 4 mm gap cuvettes. The 100 Reaction Kit includes 5 ml volume solution and one hundred 4 mm gap cuvettes.

Specifications

Specifications

  • Description:
    BTXpress solution kit
  • No. of reactions:
    40 in 4 mm gap cuvettes

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.