Sampling System, Sterile, Advanced Scientifics
4× 250 ml (8.5 oz.)
Supplier: Advanced Scientific, Inc.
Supplier #: 4MP0036
Sampling System, Sterile, Advanced Scientifics
Catalog # 89127-376
Supplier: Advanced Scientific, Inc.
Jump to:
Product Details & Documents
Polyethylene sample bags complete with 30.5 cm (12") c-flex tubing, pinch and cut devices, male luer injection sites, and 3.8 cm (1½") sanitary sampling system valve.
Documents
Certificates
Search by Lot NumberSpecifications
- Capacity:4× 250 ml (8.5 oz.)