Order Entry
Canada
Orders LinkContactUsLinkComponent
QuickConnect™ Mop Handles

QuickConnect™ Mop Handles

QuickConnect™ stainless steel handle with hanging loop, 41 cm (16")
Catalog # 89107-996
Supplier:   CONTEC, INC. MX
Supplier #:  2686
QuickConnect™ Mop Handles
Catalog # 89107-996
Supplier:  CONTEC, INC. MX
QuickConnect™ Mop Handles
QuickConnect™ Mop Handles

QuickConnect™ stainless steel handle with hanging loop, 41 cm (16")

Price
$50.69
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents

Interchangeable mop handles are available in carbon fiber, anodized aluminum, stainless steel or fiberglass.

  • Compatible with all QuickConnect™ frames
  • Multiple lengths available

Fully autoclavable QuickConnect™ mop handles and interchangeable mop head frames work together for maximum flexibility when cleaning in critical environments. Invest in one handle and the various interchangeable heads required for specific applications. Handles are available in several lengths and styles.

Telescoping handles are available in lightweight carbon fiber, anodized aluminum, durable stainless steel, or fiberglass. Choose from a variety of lengths. For hard-to-reach ceilings and walls, a carbon fiber handle is available that is lightweight and easy to maneuver. A one-piece stainless-steel handle is also available.

Specifications
  • Description:
    QuickConnect™ stainless steel handle with hanging loop
  • Length:
    41 cm (16")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.