Order Entry
Canada
Orders LinkContactUsLinkComponent
Corning® Pasteur Pipettes

Corning® Pasteur Pipettes

Disposable pasteur pipettes, Borosilicate glass, 6.5 mm, 228.6 mm (9"), Non sterile
Catalog # 89090-044
Supplier:   Corning
Supplier #:  7095D-9
Corning® Pasteur Pipettes
Catalog # 89090-044
Supplier:  Corning
Corning® Pasteur Pipettes
Corning® Pasteur Pipettes

Disposable pasteur pipettes, Borosilicate glass, 6.5 mm, 228.6 mm (9"), Non sterile

Price
$260.26
New Customer Promotional Price
Log in for availability
Case of 1,000
Product Details & Documents

Soda-lime or borosilicate glass.

  • Single use eliminates the danger of cross-contamination of valuable specimens or laboratory reagents
  • Features a constriction one inch below the top to facilitate plugging
  • Available in convenient bulk packs

These disposable Pasteur pipettes are ideal for rapid non volumetric transfer work in bacteriology, immunology, hematology and serology studies, as well as blood bank and clinical chemistry procedures.

Specifications
  • Ext.Ø:
    6.5 mm
  • Sterility:
    Non sterile
  • Description:
    Disposable pasteur pipettes
  • Length:
    228.6 mm (9")
  • Material:
    Borosilicate glass
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.