Order Entry
Canada
Orders LinkContactUsLinkComponent
Corning® Disposable Spinner Flasks, Corning

Corning® Disposable Spinner Flasks, Corning

Corning® 3 L Disposable Spinner Flask, 3 L, 45 mm, 188×259 mm, Plain
Catalog # 89089-804
Supplier:   Corning
Supplier #:  3563
Corning® Disposable Spinner Flasks, Corning
Catalog # 89089-804
Supplier:  Corning
Corning® Disposable Spinner Flasks, Corning
Corning® Disposable Spinner Flasks, Corning

Corning® 3 L Disposable Spinner Flask, 3 L, 45 mm, 188×259 mm, Plain

Price
$1,734.00
New Customer Promotional Price
Log in for availability
Case of 4
Product Details & Documents

The Corning® disposable spinner flask system comes ready-to-use with paddle and integrated magnet, eliminating the need for time-consuming assembly or cleaning and reassembly.

  • Paddle size and height is optimised for each vessel size
  • Integrated magnet provides smooth, even rotation at required speeds on slow-speed stirrers
  • Heat build-up in the vessel is reduced by means of a specially designed flange that raised the vessel off the stir-plate surface for the 125 and 500 ml flasks only

Molded from virgin polystyrene and gamma-irradiated, each spinner flask system assures a clean and sterile unit that is nonpyrogenic. No more concerns with detergent residues or contamination.

Made of USP Class VI and ISO 10993-18 polystyrene, the vessel is comparable to conventional glass spinner flasks for growth of suspension cell lines and any attachment-dependent cultures using microcarrier beads.

Specifications
  • Capacity:
    3 L
  • Description:
    Corning® 3 L Disposable Spinner Flask
  • Style:
    Plain
  • Diameter ×H:
    188×259 mm
  • Sidearm neck O.D.:
    45 mm
Accessories

Product Information

  • Catalog #
    Material
    Sterility
    Availability
    Price
  • Vent cap
    Vent cap
    89089-914
    High Density Polyethylene, orange
    Sterile
    Log in for availability
    Case of 4 - $68.85

Product Information

  • Catalog #
    Material
    Sterility
    Availability
    Price
  • Aseptic transfer cap
    Aseptic transfer cap
    89089-910
    High Density Polyethylene, orange
    Sterile
    Log in for availability
    Case of 2 - $630.70

Product Information

  • Catalog #
    Fits Flask
    Availability
    Price
  • Modified Impeller Assembly for Magnetically-Driven 36L Corning® Proculture® Spinner Flask, Reusable
    Modified Impeller Assembly for Magnetically-Driven 36L Corning® Proculture® Spinner Flask, Reusable
    89089-950
    36L Corning® Proculture® spinner flask (22877-088, 22877-098, 22877-108, 22877-132, 22877-138, 22877-142)
    Log in for availability
    Case of 1 - $1,450.00

Product Information

  • Catalog #
    Material
    Sterility
    Availability
    Price
  • Aseptic transfer cap
    Aseptic transfer cap
    89089-912
    High Density Polyethylene, orange
    Sterile
    Log in for availability
    Case of 2 - $689.35

Product Information

  • Catalog #
    Material
    Sterility
    Availability
    Price
  • Aseptic transfer cap
    Aseptic transfer cap
    89089-908
    High Density Polyethylene, orange
    Sterile
    Log in for availability
    Case of 2 - $641.75

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.