PTFE Plug Inserts, Ace Glass
PTFE Plug Inserts, Solid Plug, 1/4" NPT
Catalog # 89084-118
Product available on GSA Advantage®
Small Business Enterprise
Supplier: Ace Glass
Supplier #: 12867-04
PTFE Plug Inserts, Ace Glass
Catalog # 89084-118
Supplier: Ace Glass
Product available on GSA Advantage®
Small Business Enterprise
Jump to:
Product Details & Documents
These are PTFE inserts for PTFE reaction vessel heads.
Solid plug.
Specifications
- Description:PTFE Plug Inserts, Solid Plug
- Size:1/4" NPT
- Cat. No.:89084-118