Adjustable Jacobs Style Chuck
5/16" Motor Shaft Jacobs Chuck for Stir Shafts up to 3/8" O.D.
Catalog # 89077-784
Supplier: Ace Glass
Supplier #: 13588-04
Jump to:
Product Details & Documents
Adjustable, Jacobs Style chucks to accomodate 1/16" to 3/8" diameter stirring shafts.
- Propeller and Mixing Shafts are made of 303/304 Stainless
- Propellers are pitched 15° to create axial flow
- Adjustable Chucks should be ordered based upon the diameter of the motor shaft
Code -02 fits 1/4" diameter motor shafts, code -04 fits 5/16" diameter motor shafts.
Delivery information: Includes chuck key.
Specifications
- Description:5/16" Motor Shaft Jacobs Chuck for Stir Shafts up to 3/8" O.D.