Orders LinkContactUsLinkComponent
Adjustable Jacobs Style Chuck
Adjustable Jacobs Style Chuck
Catalog # 89077-782
Adjustable Jacobs Style Chuck
Catalog # 89077-782
Supplier:  
Adjustable Jacobs Style Chuck
Adjustable Jacobs Style Chuck
Discontinued
Product Details & Documents
Adjustable, Jacobs Style chucks to accomodate 1/16" to 3/8" diameter stirring shafts.
  • Propeller and Mixing Shafts are made of 303/304 Stainless
  • Propellers are pitched 15° to create axial flow
  • Adjustable Chucks should be ordered based upon the diameter of the motor shaft
Code -02 fits 1/4" diameter motor shafts, code -04 fits 5/16" diameter motor shafts.
Delivery information: Includes chuck key.