Orders LinkContactUsLinkComponent
Reflux Condenser, Microscale, Jacketed, Threaded
Reflux Condenser, Microscale, Jacketed, Threaded
Condenser, Reflux, Jacketed, Microscale,Threaded, 10/10 inner, [ST] 10/10 outer
Catalog # 89075-166
Supplier:   Ace Glass
Supplier #:  9567-19
Reflux Condenser, Microscale, Jacketed, Threaded
Catalog # 89075-166
Supplier:  Ace Glass
Reflux Condenser, Microscale, Jacketed, Threaded
Reflux Condenser, Microscale, Jacketed, Threaded
Condenser, Reflux, Jacketed, Microscale,Threaded, 10/10 inner, [ST] 10/10 outer
Price
$238.56
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents
This microscale, water-jacketed condenser is used in reflux experiments
The top joint is a combination cap, thread and standard taper outer joint to accommodate and seal other microscale apparatus. The O.D. of the top joint has an external 13-400 thread to facilitate an o-ring and open-top phenolic cap. The internal (or I.D.) at the top is a ground standard taper outer joint. Inner standard taper joint at bottom, with drip tip. Jacket length is approximately 80mm. Inlet and outlet water connections are three-ring serrated fittings. Hose barbs for use with 5/16-inch I.D. tubing, size A hose connection. Fabricated from borosilicate glass.
Ordering information: Supplied with open-top cap (with 9mm hole), and size -010 o-ring.
Specifications
  • Description:
    Condenser, Reflux, Jacketed, Microscale,Threaded
  • Bottom [ST] joint size:
    10/10 inner
  • Top [ST] joint size:
    [ST] 10/10 outer