Orders LinkContactUsLinkComponent
Edgeless® Mop Systems, Contec®
Edgeless® Mop Systems, Contec®
Quick-Connect Stainless Steel Mop Handle, 152.4 cm (60") Long
Catalog # 89073-978
Edgeless® Mop Systems, Contec®
Catalog # 89073-978
Supplier:  
Edgeless® Mop Systems, Contec®
Edgeless® Mop Systems, Contec®
Quick-Connect Stainless Steel Mop Handle, 152.4 cm (60") Long
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
HANDLE 59IN 1 PIECE SS Catalog #89201-652
Product 89201-652 is similar to 89073-978 but manufacturer and specifications may vary.
Product Details & Documents
Edgeless® Mops are made from strong, durable knitted polyester, eliminating contamination from breakage and deterioration common with cotton and rayon string mops. Cleanroom laundered Edgeless® Mops are made from a continuous tube of 100% polyester knitted fabric looped to form mop strands that will not shed or break.
  • Durable and chemical resistant
  • Continuous loop of mop strands provides improved cleanliness, eliminating cut edges
  • Textured large head band provides scrubbing power for difficult stains and debris
  • Ideal for use in sterile environments
Patented Textured and Long Textured Edgeless Mops feature unique ribbed, twisted strands that are more sorbent than traditional string mops. They have a textured head band that is ideal when additional abrasion resistance is required.
All mops are available sterilized by gamma radiation, and can be autoclaved. Validated sterile per the AAMI/ISO Guidelines to a 10-6 SAL. Certificates of sterility are available.
Stainless steel and polypropylene single- and multi-bucket systems are available. All mop system components and bucket systems are compatible with gamma, ETO, and autoclave sterilization, unless otherwise noted. Mop heads are laundered and packaged in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom.
Ordering information: All mop system components are ordered separately. Mop heads work well with existing wet mop hardware, but a down-press ringer is recommended for maximum extraction. Polypropylene and stainless steel bucket systems include cart, buckets, wringer, and casters.
Specifications
  • Description:
    Quick-Connect Stainless Steel Mop Handle, 152.4 cm (60") Long
Accessories
Product Information
  • Catalog #
    Material
    Capacity
    Availability
    Price
  • 20 L (5 gal.) Bucket, Stainless Steel (Replacement Part, Optional 3rd Bucket), Autoclavable
    20 L (5 gal.) Bucket, Stainless Steel (Replacement Part, Optional 3rd Bucket), Autoclavable
    33503-124
    Stainless steel
    27 L (7 galllon)
    Log in for availability
    Each - $1,659.80