Order Entry
Canada
Orders LinkContactUsLinkComponent
Condenser, Allihn, Jacketed

Condenser, Allihn, Jacketed

Condenser, Alihn, Jacketed, 200 mm
Catalog # 89053-782
Supplier:   Ace Glass
Supplier #:  5941-12
Condenser, Allihn, Jacketed
Catalog # 89053-782
Supplier:  Ace Glass
Condenser, Allihn, Jacketed
Condenser, Allihn, Jacketed

Condenser, Alihn, Jacketed, 200 mm

Price
$381.99
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents

Bulb type Allihn condenser, approximately one bulb per every 50 mm, with 24/40 standard taper inner joint at bottom, open tube at the top

Fabricated from borosilicate glass. Use with 3/8-inch I.D. tubing, size D hose connections.

Specifications
  • Description:
    Condenser, Alihn, Jacketed
  • Jacket length:
    200 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.