Order Entry
Canada
Orders LinkContactUsLinkComponent
Large Scale Evaporator Flasks, Buchi®

Large Scale Evaporator Flasks, Buchi®

20 L, R-153/R-220, Evaporating flask, 150 mm
Catalog # 89034-512
Supplier:   Buchi
Supplier #:  027468
Large Scale Evaporator Flasks, Buchi®
Catalog # 89034-512
Supplier:  Buchi
Large Scale Evaporator Flasks, Buchi®
Large Scale Evaporator Flasks, Buchi®

20 L, R-153/R-220, Evaporating flask, 150 mm

Price
$4,594.64
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Standard glass flask for use with various Rotavapor™ Evaporator models.

Specifications
  • Capacity:
    20 L
  • Description:
    Evaporating flask
  • Flange O.D.:
    150 mm
  • Fits models:
    R-153/R-220