Order Entry
Canada
Orders LinkContactUsLinkComponent
 

VWR® Jars, PTFE

355 ml (12 oz.), 95 mm, 90 mm
  89026-168
VWR® Jars, PTFE
  89026-168
VWR® Jars, PTFE
VWR® Jars, PTFE

355 ml (12 oz.), 95 mm, 90 mm

 
$650.90
 
   
Each
 

Isostatically molded, pure FDA approved PTFE molded stackable jars have thick wall construction and a smooth internal finish.

  • Chemically resistant and inert
  • Screw-on cap included

Suitable for IPA Method 1311 TCLP.

Caution: Do not use with cap sealed for chemical reactions or digestions. Loosen lid when thermally sterilizing.

 
  • Cap Size:
    90 mm
  • Capacity:
    355 ml (12 oz.)
  • Height:
    95 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.