VWR® Dropping Bottles, PTFE
25 ml (0.85 oz.), 33×80 mm
Catalog # 89026-086
Supplier: VWR International
Jump to:
Product Details & Documents
PTFE dropping bottles are suitable for dispensing and dropping small volumes of reagents and aggressive chemicals.
- Chemically resistant and inert
- Tight, leak-free seal
- Bottle sides are flexible
Bottle sides are flexible to enable user to squeeze out contents. Screw-on cap ensures a tight, leak-free seal.
Specifications
- Capacity:25 ml (0.85 oz.)
- Diameter ×H:33×80 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



