Diversified Biotech CoolSafe™ Systems
CoolSafe System for 0.2 mL Tubes (Also fits PCR Plates), For PCR Plates or 0.2 mL Tubes
Catalog # 89024-860
Supplier: Diversified Biotech
Supplier #: CSAF-1000
Jump to:
Product Details & Documents
Anodized aluminum storage kits are ideal for sustaining cool or warm constant temperatures up to 10 hours during benchtop storage
Each kit includes one polystyrene CoolSafe storage box, one anodized aluminum cooling chamber rack, and two interchangeable cool bricks. Additional aluminum cooling chamber racks are also available.
Specifications
- Capacity:For PCR Plates or 0.2 mL Tubes
- Description:CoolSafe System for 0.2 mL Tubes (Also fits PCR Plates)
Accessories
Product Information
- Catalog #DetailsAvailabilityPrice
- Replacement Cooling Chamber RacksReplacement Cooling Chamber Racks89024-880Holds Forty-Eight 1.5 mL TubesLog in for availabilityEach - $1,301.34
Product Information
- Catalog #DetailsAvailabilityPrice
- Replacement Cooling Chamber RacksReplacement Cooling Chamber Racks89024-882Holds Fifty-Five 1.5 mL Cryogenic TubesLog in for availabilityEach - $1,289.09
Product Information
- Catalog #DetailsAvailabilityPrice
- Replacement Cooling Chamber RacksReplacement Cooling Chamber Racks89024-884Holds One 384-Well Plate, Six 0.5 mL Tubes, and Four 1.5 mL TubesLog in for availabilityEach - $2,268.99
Product Information
- Catalog #DetailsAvailabilityPrice
- Replacement Cooling Chamber RacksReplacement Cooling Chamber Racks89024-878Holds Ninety-One 0.5 mL TubesLog in for availabilityEach - $1,752.18
Product Information
- Catalog #DetailsAvailabilityPrice
- Replacement Cooling Chamber RacksReplacement Cooling Chamber Racks89024-886Holds Thirty-Two 5 mL TubesLog in for availabilityEach - $1,449.35
Product Information
- Catalog #DetailsAvailabilityPrice
- Replacement Cooling Chamber RacksReplacement Cooling Chamber Racks89024-876Holds One PCR Plate or Ninety-Six 0.2 mL Tubes, Six 0.5 mL Tubes, and Four 1.5 mL TubesLog in for availabilityEach - $1,462.18
Product Information
- Catalog #DetailsAvailabilityPrice
- Replacement Cooling Chamber RacksReplacement Cooling Chamber Racks89024-888Holds Twenty-Four 0.2 mL Tubes, Thirty-Five 0.5 mL Tubes, and Twenty 1.5 mL TubesLog in for availabilityEach - $1,396.85
Product Information
- Catalog #DetailsAvailabilityPrice
- Replacement Cooling Chamber RacksReplacement Cooling Chamber Racks89024-890Holds One Flat-Bottom Microplate, Six 0.5 mL Tubes, and Four 1.5 mL TubesLog in for availabilityEach - $1,326.60