Precision Screwdriver, Aven
Phillips Driver, 50 mm (2"), No. 00
Catalog # 89012-004
Supplier #: 13911
Jump to:
Discontinued
Product Details & Documents
Specifically designed for electronics
Specifications
- Description:Phillips Driver
- Tip size:No. 00
- Blade length:50 mm (2")
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



