Protector® Demonstration Hoods, Labconco®
Product Details & Documents
Designed for classroom use, these lightweight hoods feature an epoxy-coated aluminum frame with tempered safety glass front sash, sides, rear baffle, and top.
Hood systems include a white, epoxy-coated tubular steel cart with a black 2.5cm (1") thick solid epoxy dished work surface, a handle bar, a bottom shelf, and 10.2 cm (4") diameter toe-locking casters. Also included is a FilterMate™ Portable Exhauster, which requires either two carbon filters or one carbon filter and one HEPA filter (filters sold separately).
The organic carbon filter cell is a type AC filter that contains 5.4 kg (12 lbs.) of granular activated carbon media suitable for organic solvents including acetone, ethyl acetate, toluene, and xylene. The formaldehyde carbon filter cell is a type C1 filter that contains 6.4 kg (14 lbs.) of impregnated carbon media suitable for vapors from formaldehyde. The ammonia filter cell is a type CX filter that contains 7.3 kg (16 lbs.) of impregnated carbon media suitable for ammonia and low-molecular weight amines. The HEPA filter cell is 99.99% efficient on particulates 0.3 µm in size; however, it should not be used to capture biohazardous materials or volatile organic material. The syringe kit is for carbon saturation detection (detector tubes sold separately).
Hoods have a nominal width of 91.4 cm (3') and an interior depth of 58.4 cm (23") to easily accommodate a variety of apparatus. Hoods feature the Clean-Sweep™ air foil that allows air to sweep the work surface for maximum containment; two utility ports with iris openings for connection of tubing, cords, and cables; and an ergonomic angled sash that reduces glare. Flexible exhaust options enable the hood to be connected to ductwork for exhausting to the outside or to the optional FilterMate™ Portable Exhauster (see 82010-632 series) for filtered removal of fumes and/or particulates.
Certifications: All hoods conform to ASHRAE 110-1995 standards. The 115 V models are ETL and ETL-C listed. The 230 V models are CE marked.
Ordering information: Hoods 89000-162 and 89000-104 require a connection to a remote blower, house exhaust, or FilterMate™ Portable Exhauster (see 82010-632 series). They also require a work surface and a supporting base. For Guardian airflow monitors, see 82007-280 series. For exhaust dampers, remote blowers, transition adapters for connecting ductwork, and other accessories, contact your VWR sales representative.
Specifications
- Electrical:115 V, 60 Hz, 10 A
- Shipping weight:135 lbs.
- Depth:584 mm (23")
- Width:914 mm (36")
- Enclosure height:32"
- Rediship:No
Accessories
Product Information
- Catalog #REDISHIPShipping WeightAvailabilityPrice
- Solid Epoxy Dished Work Surface for 3' ModelsSolid Epoxy Dished Work Surface for 3' Models82010-692Yes72 lbs.Log in for availabilityEach - $2,010.14
Product Information
- Catalog #REDISHIPShipping WeightAvailabilityPrice
- Fluorescent Light Kit, 115V, 60Hz, 10A
Product Information
- Catalog #REDISHIPShipping WeightAvailabilityPrice
- Fluorescent Light Kit, 230V, 50Hz, 10A
Product Information
- Catalog #REDISHIPShipping WeightAvailabilityPrice
- Cart, White Epoxy-Coated Tubular Steel
Product Information
- Catalog #Shipping WeightAvailabilityPrice
- Syringe Kit
Product Information
- Catalog #For Use WithREDISHIPAvailabilityPrice
- FilterMate™ Carbon Filter, AmmoniaFilterMate™ Carbon Filter, Ammonia82010-684Precise HEPA-Filtered Glove Boxes and XPert Weigh BoxesNoLog in for availabilityEach - $1,110.00
Product Information
- Catalog #For Use WithREDISHIPAvailabilityPrice
- Bag-In/Bag-Out Bag for HEPA FilterBag-In/Bag-Out Bag for HEPA Filter82010-688Precise HEPA-Filtered Glove Boxes and XPert Weigh BoxesNoLog in for availabilityEach - $623.00
Product Information
- Catalog #For Use WithREDISHIPAvailabilityPrice
- FilterMate™ Carbon Filter, OrganicFilterMate™ Carbon Filter, Organic82010-680Precise HEPA-Filtered Glove Boxes and XPert Weigh BoxesNoLog in for availabilityEach - $644.00
Product Information
- Catalog #For Use WithREDISHIPAvailabilityPrice
- FilterMate™ HEPA FilterFilterMate™ HEPA Filter82010-686Precise HEPA-Filtered Glove Boxes and XPert Weigh BoxesNoLog in for availabilityEach - $1,054.50
Product Information
- Catalog #Shipping WeightAvailabilityPrice
- Ammonia Detector Tubes, Set of 3
Product Information
- Catalog #For Use WithREDISHIPAvailabilityPrice
- FilterMate™ Carbon Filter, FormaldehydeFilterMate™ Carbon Filter, Formaldehyde82010-682Precise HEPA-Filtered Glove Boxes and XPert Weigh BoxesNoLog in for availabilityEach - $1,110.00
Product Information
- Catalog #Shipping WeightAvailabilityPrice
- Formaldehyde Detector Tubes, Set of 3Formaldehyde Detector Tubes, Set of 346000-2500.5 kg (1 lbs.)Log in for availabilityEach - $244.00



