Order Entry
Canada
Orders LinkContactUsLinkComponent
StatSpin® Express 3 Centrifuge, Iris Sample Processing

StatSpin® Express 3 Centrifuge, Iris Sample Processing

Centrifuge
Catalog # 87003-180
UL Listed
Supplier #:  SSX3
StatSpin® Express 3 Centrifuge, Iris Sample Processing
Catalog # 87003-180
Supplier:  
UL Listed
StatSpin® Express 3 Centrifuge, Iris Sample Processing
StatSpin® Express 3 Centrifuge, Iris Sample Processing

Centrifuge

Product Details & Documents

When rapid serum or plasma is needed, such as in coagulation testing, sample prep for stat chemistry, and cardiac profiles in the ER, this centrifuge is ideal.

  • Settings for 2, 3, and 5 minutes
  • Tubes spun in 2-minute setting equal tubes spun in 7 to 10 minute setting in other centrifuges
  • Up to 8 standard glass or plastic blood-collection tubes accepted
  • Accepted tube size: Between 1.5 to 10 ml
  • Maintenance-free, brushless motor

It has settings for 2, 3, and 5 minutes.

The centrifuge accepts up to eight standard glass or plastic blood-collection tubes from 1.5 to 10 ml. The rugged, brushless motor is maintenance free.

The two-minute setting produces serum or plasma equal to tubes spun for 7 to 10 minutes in conventional clinical centrifuges.

Certifications: UL listed. CE marked.

Ordering information: Includes 8-place rotor, removable bowl liner, universal power supply, 120 V adapter and power cord, and eight adapters each for 5 and 10 ml tubes.

Caution: Note: StatSpin centrifuges have a 6 month warranty.

Services: VWRCATALYST™ offers validation services including IQ, OQ, and PQ protocol development and execution, as well as asset management and calibration services (US only); call 888-793-2300 for more details.

Documents

Specifications
  • Description:
    Centrifuge
  • Cat. No.:
    87003-180
  • Max. speed:
    7200 rpm
  • Acceleration/braking time:
    20 s (to 95% of speed) / <30 second s
  • Dimensions:
    18H x 26.7W x 25.4D cm (71/8 x 101/2 x 10")
  • Electrical:
    120–240V, 50/60Hz, 3A
  • Max. RCF:
    4440 g ±5%
  • Cycle time:
    2, 3, 5 min
  • Weight:
    4.3 kg (9.5 lbs.)
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Inserts for BD™ Microtubes
    Inserts for BD™ Microtubes
    CA87003-962
    Log in for availability
    Pack of 4 - $327.05

Product Information

  • Catalog #
    Availability
    Price
  • Inserts for 7 mL (13 x 100 mm) Tubes
    Inserts for 7 mL (13 x 100 mm) Tubes
    CA87003-970
    Log in for availability
    Pack of 8 - $313.10

Product Information

  • Catalog #
    Availability
    Price
  • Inserts for 7 mL (16 x 75 mm) Tubes
    Inserts for 7 mL (16 x 75 mm) Tubes
    CA87003-964
    Log in for availability
    Pack of 8 - $234.70

Product Information

  • Catalog #
    Availability
    Price
  • Inserts for 5 mL (13 x 75 mm) Tubes
    Inserts for 5 mL (13 x 75 mm) Tubes
    CA87003-968
    Log in for availability
    Pack of 8 - $453.95

Product Information

  • Catalog #
    Availability
    Price
  • Disposable Bowl Liners
    Disposable Bowl Liners
    CA87003-972
    Log in for availability
    Pack of 3 - $265.76

Product Information

  • Catalog #
    Availability
    Price
  • Inserts for 10 mL (16 x 100 mm) Tubes
    Inserts for 10 mL (16 x 100 mm) Tubes
    CA87003-966
    Log in for availability
    Pack of 8 - $234.70

Product Information

  • Catalog #
    Availability
    Price
  • Rotor for 1.5–10mL Tubes, 8-Place
    Rotor for 1.5–10mL Tubes, 8-Place
    CA87003-974
    Log in for availability
    Each - $2,137.85

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.