Order Entry
Canada
Orders LinkContactUsLinkComponent
VITLAB® Forceps, PMP, BrandTech

VITLAB® Forceps, PMP, BrandTech

145 mm (5¾")
Catalog # 83009-011
Supplier:   Brandtech
Supplier #:  67995
VITLAB® Forceps, PMP, BrandTech
Catalog # 83009-011
Supplier:  Brandtech
VITLAB® Forceps, PMP, BrandTech
VITLAB® Forceps, PMP, BrandTech

145 mm (5¾")

Price
$132.95
New Customer Promotional Price
Log in for availability
Pack of 10
Product Details & Documents

Polymethylpentene (PMP) forceps with pointed ends.

  • Autoclavable at 121 °C for 20 minutes
  • Translucent

PMP is suitable for autoclaving and microwaves. Resistant to aliphatic alcohols, aldehydes, ester, aliphatic hydrocarbons, ketones, alkalis, and most acids (except oxidizing acids). Not recommended for use with ether, aromatic hydrocarbons, halogenated hydrocarbons, and oxidizing agents. Color: transparent white.

Operating temperature: 0 to 150°C (32 to 302°F)

Specifications
  • Length:
    145 mm (5¾")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.