Order Entry
Canada
Orders LinkContactUsLinkComponent
VITLAB® Forceps, PMP, BrandTech

VITLAB® Forceps, PMP, BrandTech

115 mm (4½")
  83009-010
 :   Brandtech
Supplier #:  67895
VITLAB® Forceps, PMP, BrandTech
  83009-010
 :  Brandtech
VITLAB® Forceps, PMP, BrandTech
VITLAB® Forceps, PMP, BrandTech

115 mm (4½")

 
$115.79
 
   
Pack of 10
 

Polymethylpentene (PMP) forceps with pointed ends.

  • Autoclavable at 121 °C for 20 minutes
  • Translucent

PMP is suitable for autoclaving and microwaves. Resistant to aliphatic alcohols, aldehydes, ester, aliphatic hydrocarbons, ketones, alkalis, and most acids (except oxidizing acids). Not recommended for use with ether, aromatic hydrocarbons, halogenated hydrocarbons, and oxidizing agents. Color: transparent white.

Operating temperature: 0 to 150°C (32 to 302°F)

 
  • Length:
    115 mm (4½")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.