Order Entry
Canada
Orders LinkContactUsLinkComponent
Protector® XVS Ventilation Stations, Labconco®

Protector® XVS Ventilation Stations, Labconco®

2 ft., 23", 311/4", 22¾", 29", 32", 24", 120 cfm at 80 fpm, 90 cfm at 60 fpm, No, 60 lbs.
Catalog # 82010-626
Supplier:   Labconco
Supplier #:  4862030
Protector® XVS Ventilation Stations, Labconco®
Catalog # 82010-626
Supplier:  Labconco
Protector® XVS Ventilation Stations, Labconco®
Protector® XVS Ventilation Stations, Labconco®

2 ft., 23", 311/4", 22¾", 29", 32", 24", 120 cfm at 80 fpm, 90 cfm at 60 fpm, No, 60 lbs.

Price
$7,390.00
Log in for availability
Each
Product Details & Documents

This ventilated work area is ideal as a weighing station, light-duty fume hood, science classroom workstation, solvent cleaning work bay, or for veterinary pathology and cytology

The deep 58.4cm (23") interior easily accommodates a variety of equipment including large balances. Glacier white and gray, dry powder epoxy-coated enclosures have a sturdy aluminum frame with 0.5cm (3/16") thick tempered safety glass top and side walls. The angled and hinged sash with 0.6cm (1/4") thick tempered safety glass swings up for easy loading and cleaning. The sash opening on 57.8cm (223/4") high models is 20.3cm (8"); on 81.3cm (32") high models the opening is 25.4cm (10"). The sash pivots upward and locks to a loading height of 50.8cm (20") on 57.8cm (223/4") high models, and 74.3cm (291/4") on the 81.3cm (32") high models.

The Clean-Sweep™ airfoil allows air to sweep the work surface for maximum containment. Airflow openings pull air under the airfoil, creating a constant barrier of protection from contaminants. The upper containment sash foil bleeds air into the enclosure to direct contaminants away from the operator’s breathing zone. The perforated rear baffle promotes horizontal laminar airflow to maximize containment. Additional side-entry airfoils allow air to sweep across the interior glass surfaces to enhance containment. Two utility ports allow passage of tubing and cords. Enclosures feature 5.1x25.4cm (2x10") rectangular exhaust collars on the top and bottom for connection to ductwork or portable exhauster.

Black, chemical-resistant, solid epoxy work surfaces are contoured to fit the dimensions of the enclosure and dished to contain spills. Base stands are made of durable 4.4cm (13/4") thick, tubular, glacier white epoxy-coated steel. The height of base stands is fully adjustable from 70–85cm (271/2–331/2") in 2.5cm (1") increments. Base stands come with either fixed leveling feet for use on uneven floor surfaces or four 12.7cm (5") non-marking casters.

Utility shelf kit includes three epoxy-coated steel shelves that can hold a printer, utensils, and other miscellaneous supplies. Shelves attach to the rear baffle perforations of the enclosure. Remote blowers 30153-980, 30153-982, and 82010-730 have a 1/4hp, totally enclosed, fan-cooled (TEFC) standard motor capable of overcoming external static pressure loss of 0.75in.H2O at 360cfm. Housing and wheel are made of corrosion-resistant, epoxy-coated steel. Blower inlet has an I.D. of 15.2cm (6"). Outlet dimensions are 10.8Wx18.7Lcm (41/4x73/8"). Applications involving high concentrations of solvents may require the explosion-proof blower.

Even, inward airflow protects the operator and contains hazardous or noxious fumes, vapors, and fine powders.

 : All systems conform to ASHRAE 110-1995 standards.

 : Work surface and stand are not included and must be purchased separately; contact your Avantor representative for more information. Items designated as REDISHIP can be shipped within 48 hours of receiving your order. Enclosures must be connected to either a remote blower or a FilterMate portable exhauster (see 82010-632 series) for proper exhaustion. For Guardian airflow monitors, see 82007-280 series. Gray work surfaces are also available; contact your Avantor sales representative for more information.

 : VWR CATALYST™ offers customer on-site validation, Testing & certification services including Face Velocity Profile Test, Smoke Generation Test, Tracer Gas Test, IQ, OQ, and PQ. Please call 888-793-2300 or email VWR[email protected] for more details. You can also visit https://vwr.com/equip_services.

Documents

Specifications
  • Inside height:
    311/4"
  • Inside width:
    22¾"
  • Outside height:
    32"
  • Outside depth:
    29"
  • Outside width:
    24"
  • Shipping weight:
    60 lbs.
  • Inside depth:
    23"
  • Width:
    2 ft.
  • Exhaust volume:
    120 cfm at 80 fpm, 90 cfm at 60 fpm
  • Rediship:
    No
Accessories

Product Information

  • Catalog #
    REDISHIP
    Shipping Weight
    Availability
    Price
  • Fluorescent Light Kit, 230V, 50Hz, 10A
    Fluorescent Light Kit, 230V, 50Hz, 10A
    82010-712
    No
    15 lbs.
    Log in for availability
    Each - $1,820.00

Product Information

  • Catalog #
    REDISHIP
    Shipping Weight
    Availability
    Price
  • Remote Blower, 115/230V, 50Hz, 5.6/2.8A, 35.6 x 28 x 38.1 cm (14 x 11 x 15")
    Remote Blower, 115/230V, 50Hz, 5.6/2.8A, 35.6 x 28 x 38.1 cm (14 x 11 x 15")
    30153-980
    No
    16 kg (35 lbs.)
    Log in for availability
    Each - $2,750.00

Product Information

  • Catalog #
    REDISHIP
    Shipping Weight
    Availability
    Price
  • Explosion-Proof Remote Blower, 115V, 60Hz, 4.4A
    Explosion-Proof Remote Blower, 115V, 60Hz, 4.4A
    82010-730
    No
    40 lbs.
    Log in for availability
    Each - $4,110.00

Product Information

  • Catalog #
    Electrical
    For Use With
    Availability
    Price
  • Adjustable Base Stand, Electric Hydraulic Lift
    Adjustable Base Stand, Electric Hydraulic Lift
    82003-596
    115 V, 60 Hz
    4' Models
    Log in for availability
    Each - $6,930.00

Product Information

  • Catalog #
    REDISHIP
    Shipping Weight
    Availability
    Price
  • Lower Exhaust Transition Adapter, 6" O.D. Duct
    Lower Exhaust Transition Adapter, 6" O.D. Duct
    82010-722
    Yes
    5 lbs.
    Log in for availability
    Each - $319.38

Product Information

  • Catalog #
    Electrical
    For Use With
    Availability
    Price
  • Adjustable Base Stand, Electric Hydraulic Lift
    Adjustable Base Stand, Electric Hydraulic Lift
    82003-594
    115 V, 60 Hz
    3' Models
    Log in for availability
    Each - $6,840.00

Product Information

  • Catalog #
    Outside Depth
    Outside Height
    Availability
    Price
  • Telescoping Base Stand with Fixed Feet
    Telescoping Base Stand with Fixed Feet
    89000-180
    25½"
    27½ - 33½"
    Log in for availability
    Each - $1,700.00

Product Information

  • Catalog #
    Shipping Weight
    Availability
    Price
  • Telescoping Base Stand Shelf Kit
    Telescoping Base Stand Shelf Kit
    97006-412
    9 lbs.
    Log in for availability
    Each - $237.00

Product Information

  • Catalog #
    Inside Depth
    Outside Height
    Availability
    Price
  • Solid Epoxy Dished Work Surface
    Solid Epoxy Dished Work Surface
    82010-694
    1"
    26¾"
    Log in for availability
    Each - $2,417.40

Product Information

  • Catalog #
    For Use With
    Outside Depth
    Availability
    Price
  • Adjustable Base Stand, Telescoping, with Fixed Feet
    Adjustable Base Stand, Telescoping, with Fixed Feet
    62111-176
    3' Models
    29"
    Log in for availability
    Each - $1,790.00

Product Information

  • Catalog #
    REDISHIP
    Shipping Weight
    Availability
    Price
  • Solid Epoxy Dished Work Surface for 3' Models
    Solid Epoxy Dished Work Surface for 3' Models
    82010-692
    Yes
    72 lbs.
    Log in for availability
    Each - $2,010.14

Product Information

  • Catalog #
    Outside Depth
    Outside Height
    Availability
    Price
  • Telescoping Base Stand with Casters
    Telescoping Base Stand with Casters
    82003-570
    29"
    27½ - 33½"
    Log in for availability
    Each - $2,220.00

Product Information

  • Catalog #
    Outside Depth
    Outside Height
    Availability
    Price
  • Telescoping Base Stand with Casters
    Telescoping Base Stand with Casters
    82003-572
    29"
    27½ - 33½"
    Log in for availability
    Each - $2,350.00

Product Information

  • Catalog #
    REDISHIP
    Shipping Weight
    Availability
    Price
  • Fluorescent Light Kit, 115V, 60Hz, 10A
    Fluorescent Light Kit, 115V, 60Hz, 10A
    82010-710
    No
    15 lbs.
    Log in for availability
    Each - $1,710.00

Product Information

  • Catalog #
    Availability
    Price
  • Accessory Shelf, 4'
    Accessory Shelf, 4'
    97006-416
    Log in for availability
    Each - $387.00

Product Information

  • Catalog #
    REDISHIP
    Shipping Weight
    Availability
    Price
  • Tissue Holder
    Tissue Holder
    97000-594
    No
    0.5 kg (1 lb.)
    Log in for availability
    Each - $157.00

Product Information

  • Catalog #
    For Use With
    Outside Depth
    Availability
    Price
  • Adjustable Base Stand, Telescoping, with Fixed Feet
    Adjustable Base Stand, Telescoping, with Fixed Feet
    62111-178
    4' Models
    29"
    Log in for availability
    Each - $1,890.00

Product Information

  • Catalog #
    For Use With
    Interior Dimensions
    Availability
    Price
  • Caster Kit for Hydraulic Lift Base Stands
    Caster Kit for Hydraulic Lift Base Stands
    82003-630
    Log in for availability
    Each - $325.00

Product Information

  • Catalog #
    REDISHIP
    Shipping Weight
    Availability
    Price
  • Upper Exhaust Transition Adapter, 5" I.D. Hose
    Upper Exhaust Transition Adapter, 5" I.D. Hose
    82010-716
    No
    5 lbs.
    Log in for availability
    Each - $280.00

Product Information

  • Catalog #
    REDISHIP
    Shipping Weight
    Availability
    Price
  • Bottle Holder
    Bottle Holder
    97000-592
    No
    0.5 kg (1 lb.)
    Log in for availability
    Each - $110.00

Product Information

  • Catalog #
    For Use With
    Outside Depth
    Availability
    Price
  • Adjustable Base Stand, Manual Hydraulic Lift
    Adjustable Base Stand, Manual Hydraulic Lift
    82003-582
    3' Models
    29"
    Log in for availability
    Each - $5,430.00

Product Information

  • Catalog #
    For Use With
    Interior Dimensions
    Availability
    Price
  • Seismic Support Bracket Kit, for Telescoping Base Stands with Fixed Feet
    Seismic Support Bracket Kit, for Telescoping Base Stands with Fixed Feet
    97010-022
    Log in for availability
    Each - $567.00

Product Information

  • Catalog #
    Ø
    Height
    Availability
    Price
  • Manual duct damper
    Manual duct damper
    82010-726
    15.2 cm (6")
    35.6 cm (14")
    Log in for availability
    Each - $910.00

Product Information

  • Catalog #
    REDISHIP
    Shipping Weight
    Availability
    Price
  • Utility Shelf Kit (3 Shelves)
    Utility Shelf Kit (3 Shelves)
    82010-714
    No
    2.3 kg (5 lbs.)
    Log in for availability
    Each - $268.00

Product Information

  • Catalog #
    Outside Depth
    Outside Height
    Availability
    Price
  • Telescoping Base Stand with Casters
    Telescoping Base Stand with Casters
    89000-178
    25½"
    27½ - 33½"
    Log in for availability
    Each - $2,110.00

Product Information

  • Catalog #
    Electrical
    For Use With
    Availability
    Price
  • Adjustable Base Stand, Electric Hydraulic Lift
    Adjustable Base Stand, Electric Hydraulic Lift
    82003-602
    230 V, 50 Hz
    4' Models
    Log in for availability
    Each - $6,930.00

Product Information

  • Catalog #
    REDISHIP
    Shipping Weight
    Availability
    Price
  • Remote Blower, 115V, 60Hz, 4.4A
    Remote Blower, 115V, 60Hz, 4.4A
    30153-982
    Yes
    35 lbs.
    Log in for availability
    Each - $2,460.00

Product Information

  • Catalog #
    Electrical
    For Use With
    Availability
    Price
  • Adjustable Base Stand, Electric Hydraulic Lift
    Adjustable Base Stand, Electric Hydraulic Lift
    82003-600
    230 V, 50 Hz
    3' Models
    Log in for availability
    Each - $6,840.00

Product Information

  • Catalog #
    REDISHIP
    Shipping Weight
    Availability
    Price
  • Upper Exhaust Transition Adapter, 6" O.D. Duct
    Upper Exhaust Transition Adapter, 6" O.D. Duct
    82010-718
    Yes
    5 lbs.
    Log in for availability
    Each - $319.38

Product Information

  • Catalog #
    Availability
    Price
  • Accessory Shelf, 3'
    Accessory Shelf, 3'
    97006-414
    Log in for availability
    Each - $285.00

Product Information

  • Catalog #
    For Use With
    Outside Depth
    Availability
    Price
  • Adjustable Base Stand, Manual Hydraulic Lift
    Adjustable Base Stand, Manual Hydraulic Lift
    82003-584
    4' Models
    29"
    Log in for availability
    Each - $5,510.00

Product Information

  • Catalog #
    REDISHIP
    Shipping Weight
    Availability
    Price
  • Exhaust Damper, Powder-Coated Steel,  15.2 cm (6") I.D.
    Exhaust Damper, Powder-Coated Steel,  15.2 cm (6") I.D.
    82010-724
    No
    0.5 (1)
    Log in for availability
    Each - $82.20

Product Information

  • Catalog #
    REDISHIP
    Shipping Weight
    Availability
    Price
  • Lower Exhaust Transition Adapter, 5" I.D. Hose
    Lower Exhaust Transition Adapter, 5" I.D. Hose
    82010-720
    No
    5 lbs.
    Log in for availability
    Each - $280.00