pH Electrode Solutions, WTW
KCl-250 Reference Electrolyte, 3 mol/l KCl, 250 ml
: WTW
: 109705
Solutions for operation, cleaning and storage of electrodes - simple refilling and safe storage warranted.
Electrolytes are used as refill solutions for liquid filled electrodes as well as a storage solution for all types of pH electrodes. With the pepsin cleaning solution, you can remove protein precipitation from the diaphragm of electrodes with liquid electrolyte.
- Description:KCl-250 Reference Electrolyte, 3 mol/l KCl, 250 ml