Order Entry
Canada
Orders LinkContactUsLinkComponent
pH Buffer Solutions, Color-Coded, LaMotte

pH Buffer Solutions, Color-Coded, LaMotte

500 mL, 7.00, Yellow
Catalog # 66170-852
Supplier:   LaMotte
Supplier #:  3772-L
pH Buffer Solutions, Color-Coded, LaMotte
Catalog # 66170-852
Supplier:  LaMotte
CAS Number:  
pH Buffer Solutions, Color-Coded, LaMotte

500 mL, 7.00, Yellow

CAS Number:  
Price
$49.99
Log in for availability
Each (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Minute amount of color permits immediate visual distinction of different buffer values.

Specifications

Specifications

  • Color:
    Yellow
  • Size:
    500 mL
  • pH:
    7.00

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.