Order Entry
Canada
Orders LinkContactUsLinkComponent
pH Buffer Solution, Standardized, Lamotte

pH Buffer Solution, Standardized, Lamotte

Standardized pH Buffer Solution, 500 ml, 10.00
Catalog # 66170-848
Supplier #:  2896-J
pH Buffer Solution, Standardized, Lamotte
Catalog # 66170-848
Supplier:  
CAS Number:  
pH Buffer Solution, Standardized, Lamotte

Standardized pH Buffer Solution, 500 ml, 10.00

CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
BDH BUFFER REF STD PH10 COLORLESS 500ML Catalog #BDH5078-500ML

Product BDH5078-500ML is similar to 66170-848 but manufacturer and specifications may vary.

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Standardized pH buffer solutions are used for the calibration of pH meters.

Specifications

Specifications

  • Description:
    Standardized pH Buffer Solution
  • pH:
    10.00
  • Volume:
    500 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.