Order Entry
Canada
Orders LinkContactUsLinkComponent
pH Buffer Solution, Standardized, Lamotte

pH Buffer Solution, Standardized, Lamotte

Standardized pH Buffer Solution, 500 ml, 6.86
Catalog # 66170-836
Supplier:   LaMotte
Supplier #:  2808-L
pH Buffer Solution, Standardized, Lamotte
Catalog # 66170-836
Supplier:  LaMotte
CAS Number:  
pH Buffer Solution, Standardized, Lamotte

Standardized pH Buffer Solution, 500 ml, 6.86

CAS Number:  
Price
$64.79
Log in for availability
Each (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Standardized pH buffer solutions are used for the calibration of pH meters.

Specifications

Specifications

  • Description:
    Standardized pH Buffer Solution
  • pH:
    6.86
  • Volume:
    500 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.