Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Pliers, Insertion/Extraction, Excelta

Pliers, insertion/extraction, 5 star quality, Carbon, 137.5 mm (5.5"), 4.5 mm (0.18"), 52 mm (2.080")
Catalog # 63042-454
Supplier:   Excelta
 :  505B-US
Pliers, Insertion/Extraction, Excelta
Catalog # 63042-454
Supplier:  Excelta
Pliers, Insertion/Extraction, Excelta
Pliers, Insertion/Extraction, Excelta

Pliers, insertion/extraction, 5 star quality, Carbon, 137.5 mm (5.5"), 4.5 mm (0.18"), 52 mm (2.080")

Price
$852.92
Log in for availability
Each
Product Details & Documents

These Insertion/Extraction pliers are designed to handle DIP/IC package.

  • Insertion, extraction pliers
  • Soft jaws eliminate damage to component
  • Ergonomic grips for comfort
  • Double leaf springs for ease of use

These soft tip insertion/extraction tools have soft acetal inserts eliminating scratching and nicking of delicate IC leads. The 'dash number' references the total amount of pins on package. It features a ergonomically molded antimicrobial grip (10¹⁰ ohms/sq surface resistivity) with dual leaf springs. The shanks are made of carbon tool steel.

Specifications
  • Description:
    Pliers, insertion/extraction, 5 star quality
  • Length:
    137.5 mm (5.5")
  • Material:
    Carbon
  • Jaw width:
    52 mm (2.080")
  • Jaw length:
    4.5 mm (0.18")